missing translation for 'onlineSavingsMsg'
Learn More

RHD Antibody (LFB Anti-RhD), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 30115608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30115608 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30115608 Supplier Novus Biologicals Supplier No. NBP327995JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

RHD Monoclonal antibody specifically detects RHD in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHD
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone LFB Anti-RhD
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias blood group Rh(D) polypeptide, CD240D, DIIIc, RH, Rh blood group, D antigen, RH30, Rh4, RHCED, RhDCw, RHDel, RHDVA(TT), RhII, RhK562-II, RhPI, RHPII, RHXIII
Host Species Human
Immunogen RHD/ CD240d
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6007
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.