missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHCE Polyclonal Antibody
RHCE Polyclonal Antibody
Brand: Invitrogen PA571360
This item is not returnable.
View return policy
Description
RHCE Polyclonal Antibody for Western Blot
Specifications
| RHCE | |
| Polyclonal | |
| Unconjugated | |
| RHCE | |
| (C)ces type 1 Rhesus blood group D antigen; blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; RH; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group CcEe antigens; Rh blood group protein; Rh blood group, CcEe antigens; rh polypeptide 1; Rh polypeptide I; Rh30A; Rh4; RHC; RHCE; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; RHE; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; rhesus C/E antigens; Rhesus system C and E polypeptides; RhIVb(J); RHIXB; RhPI; RhVI; RhVIII; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6006 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P18577 | |
| RHCE | |
| synthetic peptide directed towards the middle region of human RHCE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction