missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHCE Antibody (BRIC69), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65887UV
This item is not returnable.
View return policy
Description
RHCE Monoclonal specifically detects RHCE in Human samples. It is validated for Flow Cytometry.
Specifications
| RHCE | |
| Monoclonal | |
| DyLight 350 | |
| P18577 | |
| RHCE | |
| Human erythrocytes | |
| 0.1 mL | |
| Primary | |
| Recognizes human erythrocytes of normal Rh types but does not recognize Rh null erythrocytes. NB100-65887 reacts with 31kD and 35-52kD proteins which were designated CD240DCE at the 7th leucocyte typing workshop. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| BRIC69 | |
| Flow Cytometry | |
| blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6006 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction