missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561753
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: YNHKTDAALW HRSNHSNADN EFYFRYP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
RHBG encodes one of two non-erythroid members of the Rhesus (Rh) protein family. Non-erythroid Rh protein family members are mainly expressed in the kidney and belong to the methylammonium-ammonium permease/ammonia transporters superfamily. All Rh family proteins are predicted to be transmembrane proteins with 12 membrane spanning domains and intracytoplasmic N- and C-termini.Specifications
RHBG | |
Polyclonal | |
Unconjugated | |
RHBG | |
Ammonium transporter Rh type B; Rh family B glycoprotein; Rh family B glycoprotein (gene/pseudogene); rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhbg; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein; SLC42A2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H310 | |
RHBG | |
Recombinant protein corresponding to Human RHBG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only