missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHBDL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560519
This item is not returnable.
View return policy
Description
Immunogen sequence: YAVWKPQKQW ITLDTGILES PFIYSPEKRE EAWRFISY Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 32%.
RHBDL2 is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. RHBDL2 is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3.
Specifications
| RHBDL2 | |
| Polyclonal | |
| Unconjugated | |
| RHBDL2 | |
| 9130416B15; CTF; NTF; RHBDL2; rhomboid (veinlet, Drosophila)-like 2; rhomboid like 2; rhomboid protease 2; rhomboid, veinlet-like 2; rhomboid, veinlet-like 2 (Drosophila); rhomboid-like protein 2; Rhomboid-related protein 2; Rhomboid-related protein 2, C-terminal fragment; Rhomboid-related protein 2, N-terminal fragment; RRP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX52 | |
| RHBDL2 | |
| Recombinant protein corresponding to Human RHBDL2. Recombinant protein control fragment (Product #RP-100813). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction