missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDF2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23181-1-AP-150UL
This item is not returnable.
View return policy
Description
Rhomboid protease-like protein which has no protease activity but regulates the secretion of several ligands of the epidermal growth factor receptor. Indirectly activates the epidermal growth factor receptor signaling pathway and may thereby regulate sleep, cell survival, proliferation and migration.Specifications
| RHBDF2 | |
| Polyclonal | |
| Unconjugated | |
| RHBDF2 | |
| RHBDF2, RHBDL5, RHBDL6, Rhomboid 5 homolog 2, Rhomboid family member 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79651 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PJF5 | |
| RHBDF2 | |
| RHBDF2 Fusion Protein Ag19710 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction