missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RHBDD3 Polyclonal antibody specifically detects RHBDD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | RHBDD3 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C22orf3, chromosome 22 open reading frame 3, HS984G1A, pituitary tumor apoptosis, PTAG, rhomboid domain containing 3, rhomboid domain-containing protein 3 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 13 amino acid synthetic peptide from near the center of human RHBDD3. The immunogen is located within amino acids 170 - 220 of RHBDD3. Amino Acid Squence: FRWLEPSERRLQV |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?