missing translation for 'onlineSavingsMsg'
Learn More

RHBDD2 Antibody, Novus Biologicals™

Product Code. 18433491 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433491 25 μL 25µL
18475361 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18433491 Supplier Novus Biologicals Supplier No. NBP19233025ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RHBDD2 Polyclonal antibody specifically detects RHBDD2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDD2
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias H_RG122E10.2a, H_RG122E10.2b, rhomboid domain containing 2, rhomboid domain-containing protein 2, rhomboid, veinlet-like 7, veinlet-like 7 (Drosophila)
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: HMPTLPPYQPASGLCYVQNHFGPNPTSSSVYPASAGTSLGIQPPTPVNSPGTVYSGALGTPGAAGSKESSRVP
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57414
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.