missing translation for 'onlineSavingsMsg'
Learn More

RHBDD2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18429072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429072 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429072 Supplier Novus Biologicals Supplier No. NBP176249MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RHBDD2 Polyclonal antibody specifically detects RHBDD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDD2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias H_RG122E10.2a, H_RG122E10.2b, rhomboid domain containing 2, rhomboid domain-containing protein 2, rhomboid, veinlet-like 7, veinlet-like 7 (Drosophila)
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the center of human RHBDD2. The immunogen is located within amino acids 220 - 270 of RHBDD2. Amino Acid Squence: AERRAAQSRKLMPVPGSY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57414
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.