missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHBDD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520860
This item is not returnable.
View return policy
Description
A suggested positive control is K562 cell lysate. PA5-20860 can be used with blocking peptide PEP-0974.
The Rhomboid family of proteins is made up of several widely conserved polytopic membrane serine proteases that play roles in growth and development. RHBDD1 is highly expressed in the testis and is involved in the cleavage of BIK, a proapoptotic member of the Bcl-2 family. Overexpression or suppression by RNAi of RHBDD1 in 293 cells will reduce or enhance BIK-mediated apoptosis, respectively, demonstrating that RHBDD1 modulates BIK-mediated apoptotic activity. In GC-1 cells, a spermatogonia cell line that can differentiate into spermatids within the seminiferous tubules, suppression of RHBDD1 expression by RNAi caused the cells to lose the ability to survive and differentiate in mouse seminiferous tubules, suggesting that RHBDD1 may be associated with mammalian spermatogenesis.
Specifications
| RHBDD1 | |
| Polyclonal | |
| Unconjugated | |
| RHBDD1 | |
| 4930418P06Rik; HSD50; HSD-50; mRHBDD1; MSD50; MSD-50; RGD1306477; RHBDD1; Rhbdl4; rhomboid domain containing 1; rhomboid domain-containing protein 1; Rhomboid-like protein 4; rhomboid-related protein 4; rRHBDD1; RRP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 76867, 84236 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8BHC7, Q8TEB9 | |
| RHBDD1 | |
| A 14 amino acid peptide from near the carboxy terminus of human RHBDD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction