Learn More
BD Rhapsody™ Intracellular AbSeq Buffer Kit
Product Code. 32226987 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Intracellular AbSeq Buffer Kit
Unit Size:
1mL
Product Code. Product Type Unit Size
32226987 Intracellular AbSeq Buffer Kit 1mL
1 options
Product Code. 32226987 Supplier BD Supplier No. 570742
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The BD Rhapsody™ Intracellular AbSeq Buffer Kit allows users to stain for intracellular antigens using intracellular BD™ AbSeq Antibody-Oligos for profiling with the BD Rhapsody™ System.
Each intracellular BD™ AbSeq Antibody is conjugated to an antibody-specific oligonucleotide barcode for profiling alongside surface protein and mRNA expression. This product provides reagents to preserve, permeabilize, and stain cells with intracellular BD™ AbSeq Antibody-Oligos.
Prior to beginning the intracellular CITE-seq protocol, surface antigen staining should be performed using one of the following protocols:
  • BD Rhapsody™ System Single-Cell Labeling with BD™ AbSeq Ab-Oligos (1 plex to 40 plex) Protocol 23-24262 or
  • BD Rhapsody™ System Single-Cell Labeling with the BD™ Single-Cell Multiplexing Kit and BD™ AbSeq Ab-Oligos 23-21339 to support single-cell labeling with BD™ AbSeq Ab-Oligos without and with sample multiplexing, respectively
These protocols have been tested for use with up to 40 surface markers. The following protocols, BD Rhapsody™ System Single-Cell Labeling with BD™ AbSeq Ab-Oligos (41 plex to 100 plex) protocol, 23-22314 or BD Rhapsody™ System Single-Cell Labeling with BD™ Single-Cell Multiplexing Kit and BD™ AbSeq Ab-Oligos protocol, 23-22354, are available for use with high-plex experiments, however, additional optimization may be required.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Intracellular AbSeq Buffer Kit