Learn More
BD Rhapsody™ HT Xpress Package
Product Code. 18073582
Change view
Click to view available options
Product Line:
BD Rhapsody
Product Code. Product Line
18073582 BD Rhapsody
1 options
Product Code. 18073582 Supplier BD Supplier No. 666625
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The system is a higher-throughput platform that leverages BD proprietary, gentle, robust microwell-based single-cell partitioning technology to perform single cell analysis. The system is capable of single-cell capture and barcoding enabling capture of more than 640,000 cells per cartridge.
The BD Rhapsody™ HT Xpress System offers the same performance as the BD Rhapsody™ Express System, now featuring a new flexible cartridge design with the ability to run up to eight times the number of lanes at one time with a broad range of cell input, from 100–800,000 cells per cartridge.
Expanded Throughput
  • Million-cell studies now possible
  • Capable of capturing more than 640,000 cells per cartridge
  • Up to 80% cell capture rate (for certain cell types)
  • Low multiplet rate per lane
Flexible Cartridge Design
  • Up to 192 samples per cartridge (up to 24 samples per lane)
  • Partial use of cartridge enables:
  • Running more or different types of experiments
Maintain Sample Integrity
  • Gentle microwell technology
  • No sample loss due to clogging of channels
  • Recover cells with disparate size and morphology, including fragile cell types
  • Subsample and archive cell capture beads for flexibility with experimental design and collaborating across sites with equivalent data obtained from fresh beads
TRUSTED_SUSTAINABILITY
Specifications
Product Line BD Rhapsody