Learn More
BD Rhapsody™ ATAC-Seq Amplification Kit
Product Code. 32227034 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
ATAC-Seq Amplification Kit
Unit Size:
1 test
Product Code. Product Type Unit Size
32227034 ATAC-Seq Amplification Kit 1 test
1 options
Product Code. 32227034 Supplier BD Supplier No. 571356
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The BD Rhapsody™ ATAC-Seq Assays enable you to either generate highly reproducible open chromatin profiling data from single cells or perform a multiomic analysis of open chromatin accessibility and transcriptome of single cells in one experiment using the BD Rhapsody™ Single-Cell Analysis System.
Validated on the new BD Rhapsody™ Enhanced Cell Capture Beads V3, BD Rhapsody™ ATAC-Seq Assays generate consistent, high-quality data across different samples and users and a wide range of cell inputs. Key features include:
  • High sensitivity and specificity across different experimental conditions
  • Capable of accommodating a wide range of cell inputs
  • Integrated epigenomic and transcriptomic characterization on the same cells
  • Compatibility with Custom BD™ Nuclear Antibody-Oligonucleotide Conjugates
TRUSTED_SUSTAINABILITY
Specifications
Product Type ATAC-Seq Amplification Kit