missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHAMM Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA586698

Product Code. 16343565

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RHAMM
Polyclonal
Unconjugated
HMMR
AA386826; CD168; Hmmr; Hyaluronan mediated motility receptor; hyaluronan mediated motility receptor (RHAMM); hyaluronan-mediated motility receptor; hyaluronan-mediated motility receptor (RHAMM); IHABP; Intracellular hyaluronic acid-binding protein; receptor for hyaluronan-mediated motility; RHAMM
Rabbit
Antigen Affinity Chromatography
RUO
15366, 25460, 3161
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O75330, P97779, Q00547
HMMR
Synthetic peptide, corresponding to amino acids 384-429 of Human CD168.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.