missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHAMM/CD168 Antibody (S02-3E1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19597-50ul
This item is not returnable.
View return policy
Description
RHAMM/CD168 Monoclonal antibody specifically detects RHAMM/CD168 in Human samples. It is validated for Western Blot
Specifications
| RHAMM/CD168 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S02-3E1 | |
| Unconjugated | |
| CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495 | |
| A synthetic peptide of human RHAMM/CD168 (Uniprot # O75330) | |
| 50 μg | |
| Cancer, CD Markers, Cell Biology, Immunology, Stem Cells | |
| 3161 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction