missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RHAMM/CD168 Antibody (HMMR/8021) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26946PE
This item is not returnable.
View return policy
Description
RHAMM/CD168 Monoclonal antibody specifically detects RHAMM/CD168 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RHAMM/CD168 | |
| Monoclonal | |
| PE | |
| CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495 | |
| Recombinant fragment (around aa1-250) of human RHAMM/CD168 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, CD Markers, Cell Biology, Immunology, Stem Cells | |
| 3161 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| HMMR/8021 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction