missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RHAMM/CD168 Antibody (HMMR/8021) [Alexa Fluor™ 350], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26946AF350

Product Code. 30060972

  • £514.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RHAMM/CD168 Monoclonal antibody specifically detects RHAMM/CD168 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RHAMM/CD168
Monoclonal
Alexa Fluor 350
CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495
Recombinant fragment (around aa1-250) of human RHAMM/CD168 protein (exact sequence is proprietary)
0.1 mL
Cancer, CD Markers, Cell Biology, Immunology, Stem Cells
3161
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
HMMR/8021
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.