missing translation for 'onlineSavingsMsg'
Learn More

RHAMM/CD168 Antibody (2F2C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£422.00

Specifications

Antigen RHAMM/CD168
Clone 2F2C9
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18204840
View Documents
Novus Biologicals
NBP2-52437
0.1 mg
£422.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RHAMM/CD168 Monoclonal specifically detects RHAMM/CD168 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

RHAMM/CD168
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF
Unconjugated
Mouse
Human
CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495
HMMR
IgG1
Protein A or G purified
2F2C9
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Purified
RUO
PBS with 0.05% Sodium Azide
3161
Purified recombinant fragment of human CD168 (AA: 306-497 ) expressed in E. Coli.
Primary
84.1 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.