missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHAG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563110
This item is not returnable.
View return policy
Description
Immunogen sequence: ETDQTVLEQL NITKPTDMGI FFELYP Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 37%.
Associated with rhesus blood group antigen expression. May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane. {ECO:0000269; PubMed:11062476, ECO:0000269; PubMed:11861637}.
Specifications
| RHAG | |
| Polyclonal | |
| Unconjugated | |
| RHAG | |
| Ammonium transporter Rh type A; CD241; Erythrocyte membrane glycoprotein Rh50; erythrocyte plasma membrane 50 kDa glycoprotein; OHS; OHST; Rh 50 glycoprotein; rh family type A glycoprotein; rh type A glycoprotein; RH2; RH50; Rh50A; Rh50GP; RHAG; Rh-associated glycoprotein; Rhesus associated polypeptide, 50-KD; rhesus blood group family type A glycoprotein; Rhesus blood group-associated ammonia channel; Rhesus blood group-associated glycoprotein; SLC42A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02094 | |
| RHAG | |
| Recombinant protein corresponding to Human RHAG. Recombinant protein control fragment (Product #RP-101735). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction