missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHAG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551097
This item is not returnable.
View return policy
Description
RHAG Polyclonal Antibody detects endogenous levels of RHAG protein.
Associated with rhesus blood group antigen expression. May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane. {ECO:0000269; PubMed:11062476, ECO:0000269; PubMed:11861637}.
Specifications
| RHAG | |
| Polyclonal | |
| Unconjugated | |
| RHAG | |
| Ammonium transporter Rh type A; CD241; Erythrocyte membrane glycoprotein Rh50; erythrocyte plasma membrane 50 kDa glycoprotein; OHS; OHST; Rh 50 glycoprotein; rh family type A glycoprotein; rh type A glycoprotein; RH2; RH50; Rh50A; Rh50GP; RHAG; Rh-associated glycoprotein; Rhesus associated polypeptide, 50-KD; rhesus blood group family type A glycoprotein; Rhesus blood group-associated ammonia channel; Rhesus blood group-associated glycoprotein; SLC42A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6005 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| Q02094 | |
| RHAG | |
| Synthesized peptide derived from the N-terminal region of human CD241. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction