missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RHAG Polyclonal antibody specifically detects RHAG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | RHAG |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2), 40% Glycerol |
| Gene Alias | ammonium transporter Rh type A, CD241, CD241 antigen, Erythrocyte membrane glycoprotein Rh50, Erythrocyte plasma membrane 50 kDa glycoprotein, Rh family type A glycoprotein, Rh type A glycoprotein, RH50, Rh50A, RH50ARh50, Rh50GP, Rh-associated glycoprotein, Rhesus associated polypeptide, 50-KD, Rhesus blood group family type A glycoprotein, Rhesus blood group-associated ammonia channel, Rhesus blood group-associated glycoproteinRH2, SLC42A1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: ETDQTVLEQLNITKPTDMGIFFELYP |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?