missing translation for 'onlineSavingsMsg'
Learn More

RHAG Antibody, Novus Biologicals™

Product Code. 18609807 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18609807 0.1 mL 0.1mL
18630357 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18609807 Supplier Novus Biologicals Supplier No. NBP249180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RHAG Polyclonal antibody specifically detects RHAG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHAG
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias ammonium transporter Rh type A, CD241, CD241 antigen, Erythrocyte membrane glycoprotein Rh50, Erythrocyte plasma membrane 50 kDa glycoprotein, Rh family type A glycoprotein, Rh type A glycoprotein, RH50, Rh50A, RH50ARh50, Rh50GP, Rh-associated glycoprotein, Rhesus associated polypeptide, 50-KD, Rhesus blood group family type A glycoprotein, Rhesus blood group-associated ammonia channel, Rhesus blood group-associated glycoproteinRH2, SLC42A1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: ETDQTVLEQLNITKPTDMGIFFELYP
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6005
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.