missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ [Rh COD (rac)-Binap]BF4, Rh 11.2%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 439801-50-0 |
| Molecular Formula | C52H44BF4P2Rh- |
| MDL Number | MFCD16294975 |
| Quantity | 1 g |
| Synonym | r-2,2'-bis diphenylphosphino-1,1'-binaphthyl 1,5-cyclooctadiene rhodium i tetrafluoroborate, rh cod r-binap bf4, rh cod rac-binap bf4, rh cod s-binap bf4, rh cod r-binap bf4, rh, rh cod rac-binap bf4, rh, rh cod s-binap bf4, rh, inverted exclamation marka-2,2'-bis diphenylphosphino-1,1'-binaphthyl 1,5-cyclooctadiene rhodium i tetrafluoroborate, s-2,2'-bis diphenylphosphino-1,1'-binaphthyl 1,5-cyclooctadiene rhodium i tetrafluoroborate |
| InChI Key | NXRNGSUCQWUDOA-ONEVTFJLSA-N |
| SMILES | [B-](F)(F)(F)F.C1CC=CCCC=C1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Rh] |
| IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;rhodium;tetrafluoroborate |
| Molecular Weight (g/mol) | 920.581 |
| PubChem CID | 11170532 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction