missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS7BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558852
This item is not returnable.
View return policy
Description
Immunogen sequence: AEMHKTRTKG CEMARQAHQK LAAISGPEDG EIHPEICRLY IQLQCCLEMY TTEMLKSICL LGSLQFHRKG KEPGGGTKSL DCKIEESAET PAL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 70%.
Specifications
| RGS7BP | |
| Polyclonal | |
| Unconjugated | |
| RGS7BP | |
| R7 family-binding protein; R7BP; regulator of G-protein signaling 7 binding protein; regulator of G-protein signaling 7-binding protein; RGS7BP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 401190 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6MZT1 | |
| RGS7BP | |
| Recombinant protein corresponding to Human RGS7BP. Recombinant protein control fragment (Product #RP-96964). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction