missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS6 Recombinant Rabbit Monoclonal Antibody (24GB3885)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557218
This item is not returnable.
View return policy
Description
RGS6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Members of the RGS (regulator of G protein signaling) family, such as RGS6, modulate G protein function by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits (Seki et al., 1999 [PubMed 10083744]).
Specifications
| RGS6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RGS6 | |
| DKFZp313G1241; FLJ43552; GAP; H_DJ1108A12.1; MGC142132; regulator of G-protein signaling 6; regulator of G-protein signalling 6; RGS6; S914; WUGSC:H_DJ0283M22.1; WUGSC:H_DJ1108A12.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50779, 54295, 9628 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB3885 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49758, P49801, Q9Z2H2 | |
| RGS6 | |
| A synthesized peptide derived from human RGS6 (aa 403-465). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction