missing translation for 'onlineSavingsMsg'
Learn More

RGS6 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18377583 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377583 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377583 Supplier Novus Biologicals Supplier No. NBP306581AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RGS6 Polyclonal antibody specifically detects RGS6 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS6
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias DKFZp313G1241, FLJ43552, G protein signaling 6 regulator, GAP, GTPase activating protein, H_DJ0283M22.1, H_DJ1108A12.1, MGC142132, regulator of G protein signaling 6, regulator of G protein signalling 6, regulator of G-protein signaling 6, regulator of G-protein signalling 6, S914, WUGSC:H_DJ0283M22.1, WUGSC:H_DJ1108A12.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RGS6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9628
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.