Learn More
RGS5 Antibody (CL5568) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61419AF350
Product Code. 30301617
  • £558.00 / 0.10mL
Description
RGS5 Monoclonal antibody specifically detects RGS5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGS5
Monoclonal
Alexa Fluor 350
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5
This antibody was developed using a synthetic peptide derived from O15539, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
8490
Store at 4⪚C in the dark.
IgG1
Western Blot
CL5568
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified