missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS4 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E2E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66441-1-IG-20UL
This item is not returnable.
View return policy
Description
Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. This protein negatively regulates signaling upstream or at the level of the heterotrimeric G protein and is localized in the cytoplasm. Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. Regulator of G protein signaling 4 protein is 37% identical to RGS1 and 97% identical to rat Rgs4. This protein negatively regulate signaling upstream or at the level of the heterotrimeric G protein and is localized in the cytoplasm.Specifications
| RGS4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08899, P49798, P49799 | |
| RGS4 | |
| RGS4 Fusion Protein Ag6029 | |
| 20 μL | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1E2E2 | |
| Unconjugated | |
| RGS4 | |
| RGP4, RGS4, SCZD9 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100157437, 19736, 29480, 5999 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction