missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS3 Mouse anti-Human, Clone: 1D6A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66790-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the regulator of G-protein signaling (RGS) family. This protein is a GTP-ase activating protein which inhibits G-protein mediated signal transduction. The protein is largely cytosolic, but G-protein activation leads to translocation of this protein to the plasma membrane. A nuclear form of this protein has also been described, but its sequence has not been identified. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature of some transcripts is not yet known.Specifications
| RGS3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49796 | |
| RGS3 | |
| RGS3 Fusion Protein Ag3177 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1D6A3 | |
| Unconjugated | |
| RGS3 | |
| C2PA, FLJ20370, FLJ31516, FLJ90496, PDZ RGS3, RGP3, RGS3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5998 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction