missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-2564
This item is not returnable.
View return policy
Description
RGS3 Polyclonal specifically detects RGS3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RGS3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 1% BSA, 50% Glycerol with 0.02% Sodium Azide | |
| C2PA, FLJ20370, FLJ31516, FLJ90496, PDZ-RGS3, regulator of G-protein signaling 3, regulator of G-protein signalling 3, RGP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot, Immunoprecipitation | |
| P49796 | |
| RGS3 | |
| E. coli expressed human RGS 3 (amino acids 401-519). | |
| 0.1 mg | |
| Signal Transduction | |
| 5998 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction