missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RGS22 Polyclonal antibody specifically detects RGS22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Applications | Western Blot, Immunocytochemistry |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp434I092, FLJ40080, FLJ75004, MGC102908, PRTD-NY2, regulator of G-protein signaling 22, regulator of G-protein signalling 22 |
| Host Species | Rabbit |
| Purification Method | Peptide affinity purified |
| Gene ID (Entrez) | 26166 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?