missing translation for 'onlineSavingsMsg'
Learn More

RGS2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18393347 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393347 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393347 Supplier Novus Biologicals Supplier No. NBP310554100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RGS2 Polyclonal specifically detects RGS2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias Cell growth-inhibiting gene 31 protein, G0 to G1 switch regulatory 8, 24kD, G0/G1 switch regulatory protein 8, G0S8cell growth-inhibiting protein 31, regulator of G-protein signaling 2, regulator of G-protein signaling 2, 24kDa, regulator of G-protein signalling 2, 24kD, regulator of G-protein signalling 2, 24kDa
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_033087). Peptide sequence NSSAPGKPKTGKKSKQQTFIKPSPEEAQLWAEAFDELLASKYGLAAFRAF
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5997
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.