missing translation for 'onlineSavingsMsg'
Learn More

RGS19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen RGS19
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18644117
View Documents
Novus Biologicals
NBP2-62666-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18672117
View Documents
Novus Biologicals
NBP2-62666
100 μg
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RGS19 Polyclonal antibody specifically detects RGS19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

RGS19
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Autophagy, Signal Transduction
PBS (pH 7.2) and 40% Glycerol
10287
IgG
Protein A purified
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Polyclonal
Purified
RUO
Human
G alpha interacting protein, GAIPG protein signalling regulator 19, G-alpha-interacting protein, GNAI3IP, guanine nucleotide binding protein alpha inhibiting activity polypeptide 3interacting protein, regulator of G-protein signaling 19, regulator of G-protein signalling 19, RGSGAIP
This antibody was developed against a recombinant protein corresponding to amino acids: QPLPSCEVCATPSPEEVQSWAQSFDKL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.