missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583792
This item is not returnable.
View return policy
Description
Immunogen sequence: LFFSQINMCE SKEKTFFKLI HGSGKEETSK EAKIRAKEKR NRLSLLVQKP EFHEDTRSSR SGHLAKETRV SPEEAVKWGE S Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 83%.
This gene encodes a member of the regulator of G-protein signaling family. This protein is contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications
| RGS18 | |
| Polyclonal | |
| Unconjugated | |
| RGS18 | |
| regulator of G-protein signaling 18; regulator of G-protein signalling 13; regulator of G-protein signalling 18; RGS13; RGS18; RP11-142L4.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64407 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NS28 | |
| RGS18 | |
| Recombinant protein corresponding to Human RGS18. Recombinant protein control fragment (Product #RP-94313). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction