missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGS17 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584296

Product Code. 16391245

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RDSFPRFLNS QIYKSFVEST AGSSSES Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 89%.

This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGS17
Polyclonal
Unconjugated
RGS17
6430507P11Rik; hRGS17; regulator of G-protein signaling 17; regulator of G-protein signaling Z2; regulator of Gz-selective protein signaling 2; Rgs17; RGS-17; Rgsz2
Rabbit
Antigen affinity chromatography
RUO
26575
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UGC6
RGS17
Recombinant protein corresponding to Human RGS17. Recombinant protein control fragment (Product #RP-105046).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.