missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS17 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97280CL1
This item is not returnable.
View return policy
Description
RGS17 Polyclonal antibody specifically detects RGS17 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| RGS17 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| hRGS17, regulator of G-protein signaling 17, regulator of G-protein signalling 17, RGS-17, RGSZ2 | |
| Produced in rabbits immunized with E. coli-derived Human RGS17 fragment. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 26575 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction