missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS16 Antibody (OTI2C9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73884CL1
This item is not returnable.
View return policy
Description
RGS16 Monoclonal antibody specifically detects RGS16 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RGS16 | |
| Monoclonal | |
| CoraFluor 1 | |
| A28-RGS14, A28-RGS14P, hRGS-r, regulator of G-protein signaling 16, regulator of G-protein signalling 16, Retinally abundant regulator of G-protein signaling, Retinal-specific RGS, RGS-r, RGSR | |
| Full length human recombinant protein of human RGS16(NP_002919) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 6004 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2C9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction