missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11163-1-AP-20UL
This item is not returnable.
View return policy
Description
RGS10 inhibits signal transduction by increasing the GTPASE activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It associates specifically with the activated forms of the G protein subunits G(I)-ALPHA and G(Z)- alpha but fails to interact with the structurally and functionally distinct G(S)-alpha subunit. Activity on G(Z)-alpha is inhibited by palmitoylation of the G-protein.Specifications
| RGS10 | |
| Polyclonal | |
| Unconjugated | |
| RGS10 | |
| RGS10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6001, 67865 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43665, Q9CQE5 | |
| RGS10 | |
| RGS10 Fusion Protein Ag1644 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction