missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529579
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: K562. Predicted reactivity: Mouse (86%), Rhesus Monkey (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the regulator of G-protein signaling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
Specifikationer
| RGS1 | |
| Polyclonal | |
| Unconjugated | |
| RGS1 | |
| 1R20; B-cell activation protein BL34; BL34; Early response protein 1R20; epididymis secretory protein Li 87; HEL-S-87; IER1; immediate-early response 1, B-cell specific; IR20; Regulator of G-protein signaling 1; regulator of G-protein signalling 1; RGS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5996 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q08116 | |
| RGS1 | |
| Recombinant protein encompassing a sequence within the center region of human RGS1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering