missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGS1 Recombinant Rabbit Monoclonal Antibody (HL1402)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA547033

Product Code. 17819535

  • £395.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene encodes a member of the regulator of G-protein signaling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGS1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q08116, Q9JL25
RGS1
Recombinant protein encompassing a sequence within the center region of human RGS1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL1402
Unconjugated
RGS1
1R20; B-cell activation protein BL34; BL34; Early response protein 1R20; epididymis secretory protein Li 87; HEL-S-87; IER1; immediate-early response 1, B-cell specific; IR20; Regulator of G-protein signaling 1; regulator of G-protein signalling 1; RGS1
Rabbit
Protein A
RUO
50778, 5996
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.