missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93641-0.02ml
This item is not returnable.
View return policy
Description
RGR Polyclonal antibody specifically detects RGR in Rat samples. It is validated for Western Blot
Specifications
| RGR | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| retinal G protein coupled receptor, RGR-opsin, RP44, RPE retinal G-protein coupled receptor, RPE-retinal G protein-coupled receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 156-295 of human RGR (NP_002912.2). HYDYEPLGTCCTLDYSKGDRNFTSFLFTMSFFNFAMPLFITITSYSLMEQKLGKSGHLQVNTTLPARTLLLGWGPYAILYLYAVIADVTSISPKLQMVPALIAKMVPTINAINYALGNEMVCRGIWQCLSPQKREKDRTK | |
| 0.02 mL | |
| Vision | |
| 5995 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction