missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGPD5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848722
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848722 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848722 Supplier Invitrogen™ Supplier No. PA520546

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human thymus tissue lysate. PA5-20546 can be used with blocking peptide PEP-0666.

The RANBP2-like and GRIP domain containing 5 protein (RGPD5) has high similarity to RANBP2, a large RAN-binding protein localized at the cytoplasmic side of the nuclear pore complex. The gene coding for RGPD5 is thought to have arisen from a gene duplication event of RANBP2 as these highly homologous genes are located close to each other at chromosome 2q11-q12. RGPD5 was identified as an HIV dependency factor (HDF), suggesting that RGPD5 may be an important drug target in HIV treatment. At least two isoforms of RGPD5 are known to exist, of which the shorter isoform is expressed primarily in testis, while the longer of the two is expressed at low levels in a number of somatic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGPD5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RGPD5
Gene Accession No. Q99666
Gene Alias BS63; BS-63; epididymis luminal protein 161; HEL161; ran-binding protein 2-like 1; ran-binding protein 2-like 1/2; RANBP2L1; RANBP2L2; ranBP2-like 1; ranBP2-like 1/2; RANBP2-like and GRIP domain containing 5; RANBP2-like and GRIP domain-containing protein 5; RANBP2-like and GRIP domain-containing protein 5/6; RANBPL1; RGP5; RGP6; RGP7; RGPD5; RGPD6; RGPD7; Sperm membrane protein BS-63
Gene Symbols RGPD5
Host Species Rabbit
Immunogen A 13 amino acid peptide from near the amino terminus of human RGPD5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84220
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.