missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGPD5 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41084CL1
This item is not returnable.
View return policy
Description
RGPD5 Polyclonal antibody specifically detects RGPD5 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RGPD5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 84220 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| BS63, BS-63, Epididymis Luminal Protein 161, HEL161, Ran-Binding Protein 2-Like 1, Ran-Binding Protein 2-Like 1/2, ranBP2L1, RANBP2L2, RanBP2-Like 1, RanBP2-Like 1/2, RANBP2-Like And GRIP Domain Containing 5, RANBP2-Like And GRIP Domain-Containing Protein 5, RANBP2-Like And GRIP Domain-Containing Protein 5/6, RGP5, RGP6, RGP7, RGPD7, Sperm Membrane Protein BS-63 | |
| Antibody was raised against a 23 amino acid synthetic peptide from near the center of human RGPD5. The immunogen is located within amino acids 1130 - 1180 of RGPD5. Amino Acid Squence: DGDAKLERLAAKFKTPELAEEFK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction