missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGPD5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | RGPD5 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636445
|
Novus Biologicals
NBP2-41083-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18163938
|
Novus Biologicals
NBP2-41083 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGPD5 Polyclonal specifically detects RGPD5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RGPD5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 84220 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human RGPD5. The immunogen is located within amino acids 50 - 100 of RGPD5. Amino Acid Squence: LYELEENTEKAVE | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BS63, BS-63, Epididymis Luminal Protein 161, HEL161, Ran-Binding Protein 2-Like 1, Ran-Binding Protein 2-Like 1/2, ranBP2L1, RANBP2L2, RanBP2-Like 1, RanBP2-Like 1/2, RANBP2-Like And GRIP Domain Containing 5, RANBP2-Like And GRIP Domain-Containing Protein 5, RANBP2-Like And GRIP Domain-Containing Protein 5/6, RGP5, RGP6, RGP7, RGPD7, Sperm Membrane Protein BS-63 | |
| RGPD5 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title