missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGPD Pan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539396
This item is not returnable.
View return policy
Description
This antibody detects RGPD2, RGPD3, RGPD4, RGPD6 and RGPD8.
This gene is located in a cluster of Ran-binding protein related genes on chromosome 2 which arose through duplication in primates. The encoded protein contains an N-terminal TPR (tetratricopeptide repeat) domain, two Ran-binding domains, and a C-terminal GRIP domain (golgin-97, RanBP2alpha, Imh1p and p230/golgin-245) domain. [provided by RefSeq, Sep 2011].
Specifications
| RGPD pan | |
| Polyclonal | |
| Unconjugated | |
| RGPD8 | |
| RANBP2-like and GRIP domain-containing protein 2, Ran-binding protein 2-like 2, RanBP2-like 2, RanBP2L2, RGPD2, RANBP2L2, RGP2, RanBP2-like and GRIP domain-containing protein 3, RGPD3, RGP3, RanBP2-like and GRIP domain-containing protein 4, RGPD4, RGP4, RanBP2-like and GRIP domain-containing protein 8, RGPD8, RGP8, RanBP2-like and GRIP domain-containing protein 6, RGPD6, RGP6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285190, 653489, 727851, 729857, 84220 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6NKT7, O14715, P0DJD1, Q7Z3J3, Q99666 | |
| RGPD2, RGPD3, RGPD4, RGPD5, RGPD8 | |
| A synthetic peptide derived from the C-terminal region of human RGPD2/3/4/6/8 | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction