missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556056
This item is not returnable.
View return policy
Description
Immunogen sequence: ALYSLFPDHH VKKYFDQVDI SNGLDWSLDH KIFYYIDSLS YSVDAFDYDL QTGQISNRRS VYKLEKEEQI PDGMC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
The protein encoded by this gene is a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5' UTRs, but encoding the same protein.
Specifications
| RGN | |
| Polyclonal | |
| Unconjugated | |
| Rgn | |
| AI265316; epididymis secretory protein Li 41; gluconolactonase; GNL; HEL-S-41; RC; Reguc; regucalcin; regucalcin (senescence marker protein-30); Rgn; Senescence marker protein 30; Smp30; SMP-30 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9104 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15493 | |
| Rgn | |
| Recombinant protein corresponding to Human RGN. Recombinant protein control fragment (Product #RP-95632). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction