missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RGM-C/Hemojuvelin Antibody (DISC-0974) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28395-1mg
This item is not returnable.
View return policy
Description
RGM-C/Hemojuvelin Monoclonal antibody specifically detects RGM-C/Hemojuvelin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| RGM-C/Hemojuvelin | |
| Monoclonal | |
| Unconjugated | |
| Q6ZVN8 | |
| Human | |
| Protein A purified | |
| RUO | |
| 148738 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| DISC-0974 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2AMGC23953, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMCRGM domain family member C | |
| RGMC/ HFE2 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction