missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557492
This item is not returnable.
View return policy
Description
Immunogen sequence: KLLTNLPAAA VLSAQVYSAV LRGLWEENVC GTPGRTRVCT ALLYGQVCPF QDSTDGLRTI TSILFNWPPE NTSVYYQPPQ RSSFRIKLAF RNLSWPGLGL EDHQEIVLGQ LVLPEPNEAK PDDPAPRPGQ HAL Highest antigen sequence identity to the following orthologs: Mouse - 20%, Rat - 20%.
Ral Guanine Nucleotide Dissociation Stimulator-Like 4 (RGL4) is a gene that encodes a member of the RalGDS family, which is involved in the regulation of the Ras signaling pathway. This gene is located on chromosome 22 and is highly expressed in various tissues, including the brain and reproductive organs. RGL4 is known to interact with RalA and RalB, small GTPases that play crucial roles in cellular processes such as vesicular trafficking, cytoskeleton dynamics, and cell proliferation. Studies have shown that the expression of RGL4 has significant implications in various cancers. For instance, low expression of RGL4 is associated with poor prognosis and increased immune infiltration in lung adenocarcinoma (LUAD). Conversely, overexpression of RGL4 has been correlated with resistance to Midostaurin in FLT3-ITD mutant acute myeloid leukemia (AML) samples. These findings highlight the gene's potential as a biomarker for prognosis and therapy response in certain cancers, making it a valuable target for further research and clinical applications.
Specifications
| RGL4 | |
| Polyclonal | |
| Unconjugated | |
| RGL4 | |
| hRGR; ral guanine nucleotide dissociation stimulator like 4; ral guanine nucleotide dissociation stimulator-like 4; RalGDS related oncogene; Ral-GDS related protein Rgr; ralGDS-like 4; ral-GDS-related protein; RGL4; RGR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 266747 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZJ4 | |
| RGL4 | |
| Recombinant protein corresponding to Human RGL4. Recombinant protein control fragment (Product #RP-91551). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction