missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05804-50ul
This item is not returnable.
View return policy
Description
RGL4 Polyclonal antibody specifically detects RGL4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RGL4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| MGC119678, ral guanine nucleotide dissociation stimulator-like 4, RalGDS related oncogene, Ral-GDS related protein Rgr, ralGDS-like 4, ral-GDS-related protein, Rgr | |
| Produced in rabbits immunized with E. coli-derived Human RGL4 fragment. | |
| 50 μg | |
| Signal Transduction | |
| 266747 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction