missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528280
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (93%), Pig (94%), Rhesus Monkey (100%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.
Specifications
| RGL2 | |
| Polyclonal | |
| Unconjugated | |
| RGL2 | |
| GDS-related protein; HKE1.5; KE1.5; RAB2L; ral guanine nucleotide dissociation stimulator like 2; ral guanine nucleotide dissociation stimulator,-like 2; ral guanine nucleotide dissociation stimulator-like 2; RalGDS-like 2; ralGDS-like factor; Ras-associated protein RAB2L; Rgl2; Rgt2; Rlf | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O15211 | |
| RGL2 | |
| Recombinant protein encompassing a sequence within the center region of human RGL2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction